Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDPN
Entrez Gene ID
403886
Gene Name
podoplanin
Gene Aliases
GP40
Chromosome Location
Chr.2: 83552735 - 83581375 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003220.1 NP_001003220.1 5 - 6 684 81
GenBank mRNA Z81018.1 - - 684 81

View Full Product Details