Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUBG1
Entrez Gene ID
403695
Gene Name
tubulin, gamma 1
Gene Aliases
TUBG
Chromosome Location
Chr.9: 20358683 - 20363455 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003105.1 NP_001003105.1 - 79 148
GenBank mRNA AF212974.1 - - 79 148

View Full Product Details