Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QDPR
Entrez Gene ID
479082
Gene Name
quinoid dihydropteridine reductase
Gene Aliases
-
Chromosome Location
Chr.3: 62970850 - 62987512 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536229.5 XP_536229.2 3 - 4 309 117

View Full Product Details