Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UQCC1
Entrez Gene ID
608882
Gene Name
ubiquinol-cytochrome c reductase complex assembly factor 1
Gene Aliases
UQCC
Chromosome Location
Chr.24: 24285147 - 24381819 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005634970.3 XP_005635027.1 4 - 5 465 92
XM_005634971.3 XP_005635028.1 4 - 5 465 92
XM_014106999.1 XP_013962474.1 4 - 5 465 92
XM_014107000.2 XP_013962475.1 3 - 4 369 92
XM_014107002.2 XP_013962477.1 4 - 5 465 92
XM_846758.5 XP_851851.1 4 - 5 465 92

View Full Product Details