Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARPC5L
Entrez Gene ID
612856
Gene Name
actin related protein 2/3 complex subunit 5 like
Gene Aliases
-
Chromosome Location
Chr.9: 58142001 - 58149285 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_858438.5 XP_863531.2 3 - 4 528 148

View Full Product Details