Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA11
Entrez Gene ID
476735
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
Gene Aliases
-
Chromosome Location
Chr.20: 54183584 - 54189435 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_533939.6 XP_533939.2 3 - 4 393 115

View Full Product Details