Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SVIP
Entrez Gene ID
100500800
Gene Name
small VCP interacting protein
Gene Aliases
-
Chromosome Location
Chr.21: 44408467 - 44418160 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195609.1 NP_001182538.1 2 - 3 332 131
GenBank mRNA DN422453.1 - - 332 131

View Full Product Details