Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMAT5
Entrez Gene ID
477536
Gene Name
zinc finger matrin-type 5
Gene Aliases
-
Chromosome Location
Chr.26: 22926327 - 22953189 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014108059.2 XP_013963534.1 5 - 6 600 92
XM_022410308.1 XP_022266016.1 6 - 7 771 92
XM_022410309.1 XP_022266017.1 5 - 6 502 92
XM_534730.6 XP_534730.1 5 - 6 551 92

View Full Product Details