Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1R
Entrez Gene ID
477707
Gene Name
complement C1r
Gene Aliases
-
Chromosome Location
Chr.27: 37951137 - 37960687 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534901.6 XP_534901.2 5 - 6 938 60

View Full Product Details