Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MZT2B
Entrez Gene ID
477569
Gene Name
mitotic spindle organizing protein 2B
Gene Aliases
-
Chromosome Location
Chr.26: 30420222 - 30427393 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_534764.5 XP_534764.2 1 - 2 205 100

View Full Product Details