Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM50A
Entrez Gene ID
478181
Gene Name
transmembrane protein 50A
Gene Aliases
-
Chromosome Location
Chr.2: 74504651 - 74523178 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005617784.3 XP_005617841.1 4 - 5 516 77
XM_535354.6 XP_535354.1 4 - 5 449 77

View Full Product Details