Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSL24D1
Entrez Gene ID
478313
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.30: 20610526 - 20628202 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_535488.6 XP_535488.2 3 - 4 429 76

View Full Product Details