Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL24
Entrez Gene ID
478547
Gene Name
ribosomal protein L24
Gene Aliases
-
Chromosome Location
Chr.33: 8018183 - 8023779 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005639430.3 XP_005639487.1 2 - 2 457 100

View Full Product Details