Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSR4
Entrez Gene ID
481082
Gene Name
signal sequence receptor subunit 4
Gene Aliases
-
Chromosome Location
Chr.X: 121608802 - 121613796 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204756.1 NP_001191685.1 4 - 5 409 123
XM_005640995.3 XP_005641052.1 5 - 6 770 123
GenBank mRNA DN393206.1 - 4 - 5 409 123

View Full Product Details