Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPM4
Entrez Gene ID
609879
Gene Name
tropomyosin 4
Gene Aliases
-
Chromosome Location
Chr.20: 46354808 - 46376489 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003432905.2 XP_003432953.1 6 - 7 776 122
XM_003432906.4 XP_003432954.1 6 - 7 715 122
XM_847548.5 XP_852641.1 7 - 8 761 122
XM_861034.5 XP_866127.1 7 - 8 767 122
GenBank mRNA KR822489.1 - 7 - 8 703 122

View Full Product Details