Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPM4
Entrez Gene ID
609879
Gene Name
tropomyosin 4
Gene Aliases
-
Chromosome Location
Chr.20: 46354808 - 46376489 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003432905.2 XP_003432953.1 7 - 8 851 97
XM_847548.5 XP_852641.1 8 - 9 836 97

View Full Product Details