Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YY1
Entrez Gene ID
611171
Gene Name
YY1 transcription factor
Gene Aliases
-
Chromosome Location
Chr.8: 68539511 - 68572796 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_849421.5 XP_854514.1 3 - 4 983 124

View Full Product Details