Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB1
Entrez Gene ID
476688
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
Gene Aliases
-
Chromosome Location
Chr.20: 48120604 - 48131091 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_847807.5 XP_852900.1 1 - 2 300 101

View Full Product Details