Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAU
Entrez Gene ID
483748
Gene Name
FAU ubiquitin like and ribosomal protein S30 fusion
Gene Aliases
-
Chromosome Location
Chr.18: 52072705 - 52074337 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005631491.3 XP_005631548.1 2 - 3 226 113
XM_022405677.1 XP_022261385.1 2 - 3 281 113

View Full Product Details