Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGSN
Entrez Gene ID
481862
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
GLULD1, LGS
Chromosome Location
Chr.12: 26852832 - 26884307 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048103.1 NP_001041568.1 3 - 4 379 83
XM_014118049.2 XP_013973524.1 3 - 4 469 83
GenBank mRNA DQ415892.1 - 3 - 4 379 83

View Full Product Details