Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRAGD
Entrez Gene ID
474994
Gene Name
Ras related GTP binding D
Gene Aliases
-
Chromosome Location
Chr.12: 48583350 - 48625032 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426350.1 XP_022282058.1 3 - 4 613 115

View Full Product Details