Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC474791
Entrez Gene ID
474791
Gene Name
protein NipSnap homolog 3A
Gene Aliases
NIPSNAP3A
Chromosome Location
Chr.11: 60715223 - 60729186 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_532021.6 XP_532021.3 4 - 5 716 132

View Full Product Details