Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPY
Entrez Gene ID
475257
Gene Name
neuropeptide Y
Gene Aliases
-
Chromosome Location
Chr.14: 37824579 - 37831289 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628637.3 XP_005628694.1 1 - 2 180 66

View Full Product Details