Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB9
Entrez Gene ID
475286
Gene Name
DnaJ heat shock protein family (Hsp40) member B9
Gene Aliases
-
Chromosome Location
Chr.14: 48879963 - 48885616 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003431935.4 XP_003431983.1 2 - 3 320 106
XM_005628728.3 XP_005628785.1 2 - 3 365 106
XM_532518.6 XP_532518.2 2 - 3 366 106

View Full Product Details