Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZK1IP1
Entrez Gene ID
475365
Gene Name
PDZK1 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 13479519 - 13486628 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014119709.2 XP_013975184.1 2 - 3 396 83

View Full Product Details