Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB5
Entrez Gene ID
476177
Gene Name
serpin family B member 5
Gene Aliases
-
Chromosome Location
Chr.1: 13566982 - 13638839 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005615344.3 XP_005615401.1 - 296 122
XM_022414958.1 XP_022270666.1 - 616 122
XM_022415006.1 XP_022270714.1 - 391 122
XM_533382.5 XP_533382.3 - 538 122

View Full Product Details