Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC9
Entrez Gene ID
474557
Gene Name
thioredoxin domain containing 9
Gene Aliases
-
Chromosome Location
Chr.10: 43343703 - 43356463 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531785.6 XP_531785.2 4 - 5 735 79

View Full Product Details