Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT2
Entrez Gene ID
474746
Gene Name
nudix hydrolase 2
Gene Aliases
-
Chromosome Location
Chr.11: 51049992 - 51064317 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531977.6 XP_531977.1 1 - 2 136 53

View Full Product Details