Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS26
Entrez Gene ID
474395
Gene Name
ribosomal protein S26
Gene Aliases
-
Chromosome Location
Chr.10: 362699 - 364406 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_531628.5 XP_531628.1 - 438 68

View Full Product Details