Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACYP2
Entrez Gene ID
474595
Gene Name
acylphosphatase 2
Gene Aliases
-
Chromosome Location
Chr.10: 54978903 - 55227085 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014117533.2 XP_013973008.1 2 - 3 256 67
XM_022424732.1 XP_022280440.1 2 - 3 264 67
XM_022424733.1 XP_022280441.1 2 - 3 265 67
XM_022424743.1 XP_022280451.1 2 - 3 271 67
XM_022424745.1 XP_022280453.1 2 - 3 261 67
XM_022424746.1 XP_022280454.1 2 - 3 260 67
XM_022424747.1 XP_022280455.1 2 - 3 259 67

View Full Product Details