Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL24
Entrez Gene ID
612293
Gene Name
methyltransferase like 24
Gene Aliases
C12H6orf186
Chromosome Location
Chr.12: 66957081 - 67104380 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022426448.1 XP_022282156.1 2 - 3 220 65

View Full Product Details