Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYROBP
Entrez Gene ID
476477
Gene Name
transmembrane immune signaling adaptor TYROBP
Gene Aliases
-
Chromosome Location
Chr.1: 116757271 - 116760453 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197117.1 NP_001184046.1 2 - 3 171 66
XM_005615749.3 XP_005615806.1 2 - 3 189 66
GenBank mRNA CO655102.1 - 3 - 4 420 66
CO704168.1 - 3 - 4 421 66
DN439202.1 - 2 - 3 163 66
FJ687210.1 - 2 - 3 88 66

View Full Product Details