Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIPR1
Entrez Gene ID
475646
Gene Name
EARP complex and GARP complex interacting protein 1
Gene Aliases
TSSC1
Chromosome Location
Chr.17: 1869924 - 1989856 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_014120475.2 XP_013975950.1 - 527 70
XM_022404403.1 XP_022260111.1 - 566 70
XM_022404404.1 XP_022260112.1 - 344 70
XM_022404405.1 XP_022260113.1 - 354 70
XM_022404406.1 XP_022260114.1 - 367 70
XM_022404407.1 XP_022260115.1 - 281 70
XM_022404408.1 XP_022260116.1 - 244 70
XM_022404409.1 XP_022260117.1 - 387 70
XM_532856.7 XP_532856.3 - 650 70

View Full Product Details