Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCVRN
Entrez Gene ID
489500
Gene Name
recoverin
Gene Aliases
RCV1
Chromosome Location
Chr.5: 34323580 - 34330667 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014281.1 NP_001014303.1 1 - 2 647 65

View Full Product Details