Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1
Entrez Gene ID
474452
Gene Name
GLI pathogenesis related 1
Gene Aliases
-
Chromosome Location
Chr.10: 15958923 - 15977013 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005625646.2 XP_005625703.1 - 677 62
XM_005625647.3 XP_005625704.1 - 889 62
XM_531682.6 XP_531682.1 - 878 62

View Full Product Details