Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TERB1
Entrez Gene ID
479692
Gene Name
telomere repeat binding bouquet formation protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 82486920 - 82537347 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005620870.3 XP_005620927.1 4 - 5 627 96
XM_005620872.3 XP_005620929.1 4 - 5 628 96
XM_005620873.2 XP_005620930.1 4 - 5 627 96
XM_005620874.2 XP_005620931.1 4 - 5 627 96
XM_014113980.2 XP_013969455.1 4 - 5 629 96
XM_014113981.2 XP_013969456.1 4 - 5 450 96
XM_014113982.2 XP_013969457.1 3 - 4 572 96
XM_022419397.1 XP_022275105.1 4 - 5 467 96
XM_022419398.1 XP_022275106.1 4 - 5 396 96
XM_022419399.1 XP_022275107.1 3 - 4 358 96
XM_022419401.1 XP_022275109.1 4 - 5 627 96

View Full Product Details