Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL25
Entrez Gene ID
480252
Gene Name
interleukin 25
Gene Aliases
-
Chromosome Location
Chr.8: 3621268 - 3624689 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005623236.3 XP_005623293.1 1 - 2 135 59

View Full Product Details