Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN2
Entrez Gene ID
481078
Gene Name
centrin 2
Gene Aliases
-
Chromosome Location
Chr.X: 120714676 - 120720012 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005641964.3 XP_005642021.1 3 - 4 443 127

View Full Product Details