Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL16
Entrez Gene ID
480604
Gene Name
C-C motif chemokine ligand 16
Gene Aliases
-
Chromosome Location
Chr.9: 37787658 - 37791132 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537724.6 XP_537724.3 1 - 2 169 58

View Full Product Details