Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS14
Entrez Gene ID
480061
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
-
Chromosome Location
Chr.7: 24342371 - 24368519 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537182.6 XP_537182.1 1 - 2 136 65

View Full Product Details