Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTRB2
Entrez Gene ID
479650
Gene Name
chymotrypsinogen B2
Gene Aliases
-
Chromosome Location
Chr.5: 75514065 - 75518690 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536782.5 XP_536782.2 1 - 2 111 71

View Full Product Details