Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GFER
Entrez Gene ID
479885
Gene Name
growth factor, augmenter of liver regeneration
Gene Aliases
-
Chromosome Location
Chr.6: 38948927 - 38951300 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537010.6 XP_537010.5 1 - 2 319 79

View Full Product Details