Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUDT16L1
Entrez Gene ID
479862
Gene Name
nudix hydrolase 16 like 1
Gene Aliases
-
Chromosome Location
Chr.6: 36711351 - 36713931 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_536987.6 XP_536987.2 1 - 2 190 74

View Full Product Details