Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERH
Entrez Gene ID
480371
Gene Name
ERH mRNA splicing and mitosis factor
Gene Aliases
-
Chromosome Location
Chr.8: 43211168 - 43225307 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_537493.6 XP_537493.4 1 - 2 174 72

View Full Product Details