Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEOX2
Entrez Gene ID
482329
Gene Name
mesenchyme homeobox 2
Gene Aliases
-
Chromosome Location
Chr.14: 30258737 - 30324438 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022427424.1 XP_022283132.1 1 - 2 953 74

View Full Product Details