Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD79A
Entrez Gene ID
484483
Gene Name
CD79a molecule
Gene Aliases
IG-alpha
Chromosome Location
Chr.1: 112411281 - 112414827 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313834.1 NP_001300763.1 4 - 5 681 82

View Full Product Details