Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AATK
Entrez Gene ID
483358
Gene Name
apoptosis associated tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.9: 911461 - 923508 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_022423892.1 XP_022279600.1 - 145 80

View Full Product Details