Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THAP5
Entrez Gene ID
482404
Gene Name
THAP domain containing 5
Gene Aliases
-
Chromosome Location
Chr.14: 48738882 - 48880295 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005628732.3 XP_005628789.1 1 - 2 208 126
XM_005628733.3 XP_005628790.1 1 - 2 208 126
XM_022427583.1 XP_022283291.1 1 - 2 208 126
XM_539521.6 XP_539521.2 1 - 2 201 126

View Full Product Details