Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC86
Entrez Gene ID
483806
Gene Name
coiled-coil domain containing 86
Gene Aliases
-
Chromosome Location
Chr.18: 55431768 - 55437612 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_540927.4 XP_540927.1 3 - 4 840 58

View Full Product Details