Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEUROD6
Entrez Gene ID
482387
Gene Name
neuronal differentiation 6
Gene Aliases
ATH2
Chromosome Location
Chr.14: 43913351 - 43916818 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_539504.6 XP_539504.1 1 - 2 262 119

View Full Product Details